Cynomolgus monkey recombinant TIM-3 (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Tag sequence:His Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilised Cynomolgus TIM-3, His Tag (Cat. No. TI3-C52H4) at 1 μg/ml (100 μl/well) can bind Anti-TIM-3 MAb with a linear range of 0,8 - 6 ng/ml (QC tested).
- Protein synonyms:TIMD3|FLJ14428|KIM3|TIM3|HAVCR2
- Protein/peptide name:TIM-3
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:21,6 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together