Order Entry
Belgium
ContactUsLinkComponent
 
Mouse recombinant IL-33 (from HEK293 cells)
 
Mouse recombinant IL-33 (from HEK293 cells)

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Tag sequence:
    His Tag (MALS verified)
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Gene ID:
    Q8BVZ5-1
  • Protein synonyms:
    DV27|NFEHEV|IL33|IL1F11|DKFZp586H0523|DVS27|NFHEV|RP11-575C20.2|C9ORF26
  • Protein/peptide name:
    IL-33
  • Purity:
    >90% as determined by SDS-PAGE
  • Molecular weight:
    19,5 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4 . Normally trehalose is added as protectant before lyophilisation
  • Shipping temperature:
    RT

 

Frequently Bought Together