Mouse recombinant IL-2 R alpha (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Mouse IL-2 R alpha / CD25 Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Tag sequence:His Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilised Biotinylated Human IL-2, Fc Tag, Avi Tag (Cat. No. IL2-H82F3) at 5 μg/ml (100 μl/well) on streptavidin precoated (0,5 μg/well) plate, can bind Mouse IL-2 R alpha, His Tag (Cat. No. ILA-M52H9) with a linear range of 5 - 40 ng/ml (QC tested). Measured by its binding ability in a functional ELISA. Immobilised Mouse IL-2 R alpha, His Tag (Cat. No. ILA-M52H9) at 5 μg/ml (100 μl/well) can bind Biotinylated Human IL-2, Fc Tag, Avi Tag (Cat. No. IL2-H82F3) with a linear range of 0,6 - 10 ng/ml (Routinely tested).
- Gene ID:P01590-1
- Protein synonyms:IL2-RA|p55|IL-2-RA|CD25|IL2RA
- Protein/peptide name:IL-2 R alpha
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:26,6 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
- Shipping temperature:RT
Specifications
Frequently Bought Together