Order Entry
Belgium
ContactUsLinkComponent
Mouse recombinant Flt-3 (from HEK293 cells)
Mouse recombinant Flt-3 (from HEK293 cells)
Supplier:  ACRO Biosystems
Certificates
Mouse recombinant Flt-3 (from HEK293 cells)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Protein synonyms:
    FLT-3|Flk-2|STK-1|Flt-3|CD135|FLK2
  • Protein/peptide name:
    Flt-3
  • Purity:
    >90% as determined by SDS-PAGE
  • Molecular weight:
    59,9 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together