Order Entry
Belgium
ContactUsLinkComponent
Human recombinant integrin alpha 2b beta 3 (from HEK293)
Human recombinant integrin alpha 2b beta 3 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant integrin alpha 2b beta 3 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human Fibronectin at 2 μg/ml (100 μl/well) can bind Human ITGAIIb&ITGB3 Heterodimer Protein, His Tag and Tag Free (Cat. No. ACRBIT3-H52W8) with a linear range of 2 - 78 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised Human ITGAIIb and ITGB3 Heterodimer Protein, His Tag and Tag Free (Cat. No. ACRBIT3-H52W8) at 1 μg/ml (100 μl/well) can bind Abciximab with a linear range of 0,8 - 6 ng/ml (Routinely tested).
  • Gene ID:
    P08514-1 (R887L) (ITGAIIb) and P05106-1 (ITGB3)
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    ITGAIIb&ITGB3|Integrin alpha 2b beta 3
  • Protein/peptide name:
    Integrin alpha 2b beta 3
  • Purity:
    >90%
  • Molecular weight:
    111,4 kDa (ITGAIIb) and 81,8 kDa (ITGB3)
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together