Human recombinant CTLA-4 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,1 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human B7-1, Fc Tag (Cat. No. ACRBB71-H5259) at 2 μg/ml (100 μl/well) can bind Human CTLA-4, mouse IgG2a Fc tag, low endotoxin (Cat. No. ACRBCT4-H52A4) with a linear range of 0,1 - 2 ng/ml (QC tested). Measured by its binding ability in a functional ELISA. Immobilised Human B7-2, Fc Tag (Cat. No. ACRBCD6-H5257) at 2 μg/ml (100 μl/well) can bind Human CTLA-4, mouse IgG2a Fc tag, low endotoxin (Cat. No. ACRBCT4-H52A4) with a linear range of 0,5 - 7,8 ng/ml (Routinely tested). Measured by its binding ability in a functional ELISA. Immobilised Ipilimumab at 5 μg/ml (100 μl/well) can bind Human CTLA-4, mouse IgG2a Fc tag, low endotoxin (Cat. No. ACRBCT4-H52A4) with a linear range of 0,1 - 0,8 ng/ml (Routinely tested). Loaded Human CTLA-4, mouse IgG2a Fc tag, low endotoxin (Cat. No. ACRBCT4-H52A4) on Protein A Biosensor, can bind Human B7-1, His Tag (Cat. No. ACRBB71-H5228) with an affinity constant of 0,52 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Human CTLA-4, mouse IgG2a Fc tag, low endotoxin (Cat. No. ACRBCT4-H52A4) on Protein A Biosensor, can bind Human B7-2, His Tag (Cat. No. ACRBCD6-H5223) with an affinity constant of 1,6 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P16410-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CTLA4|CD152
- Protein/peptide name:CTLA-4
- Purity:>95%
- Molecular weight:40,6 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together