Order Entry
Belgium
ContactUsLinkComponent
Human recombinant CD48 (from HEK293)
Human recombinant CD48 (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human recombinant CD48 (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human 2B4, His Tag (Cat. No. 2B4-H5224) at 2 μg/ml (100 μl/well) can bind Human CD48, Fc Tag (Cat. No. BC1-H5255) with a linear range of 0,078 - 2,5 μg/ml (QC tested). Measured by its binding ability in a functional ELISA. Immobilised Biotinylated Human 2B4, Avitag,His Tag (recommended for biopanning) (Cat. No. 2B4-H82E9) at 1 μg/ml (100 μl/well) on streptavidin precoated (0,2 μg/well) plate, can bind Human CD48, Fc Tag (Cat. No. BC1-H5255) with a linear range of 0,3 - 5 ng/ml (Routinely tested).
  • Gene ID:
    P09326-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    CD48|BCM-1|MEM-102|BLAST|SLAMF-2|BLAST1|BCM1|SLAMF2|BLAST-1|TCT.1
  • Protein/peptide name:
    CD48
  • Purity:
    >95%
  • Molecular weight:
    49,0 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder, RT

Specifications

Frequently Bought Together