Order Entry
Belgium
ContactUsLinkComponent
Human recombinant CD40 (from HEK293)
Human recombinant CD40 (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human recombinant CD40 (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. FACS analysis shows that Biotinylated Human CD40, Fc,Avitag (Cat. No. TN5-H82F9) can bind to 293T cells overexpressing human CD40L. The concentration of Biotinylated Human CD40 is 0,1 μg/ml (Routinely tested).
  • Gene ID:
    P25942-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    p50|CD40|MGC9013|Bp50|TNFRSF5|CDW40
  • Protein/peptide name:
    CD40
  • Purity:
    >95%
  • Molecular weight:
    48,1 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder, RT

Specifications

Frequently Bought Together