Human recombinant CD27 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human CD27, His Tag (Cat. No. CD7-H522b) at 2 μg/ml (100 μl/well) can bind Biotinylated Human CD27 Ligand, Avitag,Fc Tag (Cat. No. TN7-H82F4) with a linear range of 2 - 31 ng/ml (QC tested).
- Gene ID:P26842-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:Tp55|S152|T14|TNFRSF7|CD27
- Protein/peptide name:CD27
- Purity:>90%
- Molecular weight:21,1 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder, RT
Specifications
Frequently Bought Together