Order Entry
Belgium
ContactUsLinkComponent
Human recombinant CD229 (from HEK293)
Human recombinant CD229 (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human recombinant CD229 (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Gene ID:
    Q9HBG7-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    LY9|hly9|Ly-9|SLAMF3|Ly9
  • Protein/peptide name:
    CD229
  • Purity:
    >90%
  • Molecular weight:
    46,7 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder, RT

Specifications

Frequently Bought Together