Human recombinant NTB-A (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4, Normally trehalose is added as protectant before Lyophilisation.
- Gene ID:Q96DU3-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:SLAMF6|NK-T-B-antigen|CD352|KALI|NTB-A|Ly108
- Protein/peptide name:NTB-A
- Purity:>95%
- Molecular weight:25,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4, Normally trehalose is added as protectant before Lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together