Human recombinant Mesothelin (from HEK293)
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C, avoid light
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Anti-Human MSLN MAb at 10 μg/ml(100 μl/well) can bind GFP Fusion Human Mesothelin (296-580), His Tag (Cat. No. MSN-HG2P4) with a linear range of 0.039-0.625 μg/ml(QC tested).; Anti-human MSLN MAb (chimeric mouse-human IgG1) captured on CM5 chip via Anti-human IgG Fc antibodies surface, can bind GFP Fusion Human Mesothelin (296-580), His Tag (Cat. No. MSN-HG2P4) with an affinity constant of 0.542 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; 2e5 of the MSLN-CAR-293 cells were stained with 100 μl of 3 μg/mlof GFP Fusion Human Mesothelin (296-580), His Tag (Cat. No. MSN-HG2P4). GFP was used as negative control (Routinely tested).
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:MPF|MSLN|Mesothelin
- Protein/peptide name:Mesothelin
- Purity:>90%
- Molecular weight:60,4 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT, Avoid light
Frequently Bought Together