Human recombinant Mesothelin (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C, avoid light
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4, Normally trehalose is added as protectant before Lyophilisation,; Measured by its binding ability in a functional ELISA, Immobilised Anti-Human MSLN MAb at 10 μg/ml (100 μl/well) can bind FITC-Labeled Human Mesothelin (296-580), Fc Tag (Cat, No, MSN-HF253) with a linear range of 0,078-0,625 μg/ml (Ex,488 nm/Em,535 nm) (QC tested),; 293 cells were transfected with anti-MSLN-scFv and RFP tag, 2e5 of the cells were stained with B, FITC-Labeled Human Mesothelin (296-580), Fc Tag (Cat, No, MSN-HF253, 1 μg/ml) and C, FITC-labeled protein control, A, Non-transfected 293 cells and C, FITC-labeled protein control were used as negative control, RFP was used to evaluate CAR (anti-MSLN-scFv) expression and FITC was used to evaluate the binding activity of FITC-Labeled Human Mesothelin (296-580), Fc Tag (Cat, No, MSN-HF253) (QC tested).
- Gene ID:Q13421-3 (R309P)
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:MPF|MSLN|Mesothelin
- Protein/peptide name:Mesothelin
- Purity:>95%
- Molecular weight:58,6 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4, Normally trehalose is added as protectant before Lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT, Avoid light
Specifications
Frequently Bought Together