Order Entry
Belgium
ContactUsLinkComponent
Human recombinant M-CSF (from HEK293)
Human recombinant M-CSF (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human recombinant M-CSF (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Human M-CSF, His Tag (Cat. No. MCF-H5247) at 5 μg/ml(100 μl/well) can bind Human M-CSF R, Fc Tag (Cat. No. CSR-H5258) with a linear range of 0.1-4 ng/ml(QC tested).; Measured by its binding ability in a functional ELISA. Immobilised Human M-CSF, His Tag (Cat. No. MCF-H5247) at 5 μg/ml(100 μl/well) can bind Cynomolgus M-CSF R, Fc Tag (Cat. No. CSR-C5252) with a linear range of 1-39 ng/ml(Routinely tested).; Human M-CSF, His Tag (Cat. No. MCF-H5247) captured on CM5 Chip via anti-His antibody can bind Human M-CSF R, Fc Tag, low endotoxin (Cat. No. CSR-H5258) with an affinity constant of 1.9 nM as determined in SPR assay (Biacore T200) (Routinely tested).; Loaded Human M-CSF, His Tag (Cat. No. MCF-H5247) on HIS1K Biosensor, can bind Human M-CSF R, Fc Tag, low endotoxin (Cat. No. CSR-H5258) with an affinity constant of 3.29 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Human M-CSF R, Fc Tag, low endotoxin (Cat. No. CSR-H5258) on Protein A Biosensor, can bind Human M-CSF, His Tag (Cat. No. MCF-H5247) with an affinity constant of 0.365 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    P09603-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    CSF-1|M-CSF|Lanimostim
  • Protein/peptide name:
    M-CSF
  • Purity:
    >95%
  • Molecular weight:
    26,0 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together