Order Entry
Belgium
ContactUsLinkComponent
Human recombinant LILRB3 (from HEK293)
Human recombinant LILRB3 (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human recombinant LILRB3 (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before Lyophilisation.
  • Gene ID:
    NP_006855.2
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    ILT5|LILRB3|CD85a
  • Protein/peptide name:
    LILRB3
  • Purity:
    >95%
  • Molecular weight:
    74,6 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before Lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together