Human recombinant LILRA2 (from HEK293)
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) at 5 μg/ml(100 μl/well) can bind Biotinylated Human LILRA2, His,Avitag (Cat. No. LI2-H82E9) with a linear range of 0.625-5 μg/ml(QC tested).
- Gene ID:Q8N149-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:ILT1CD85H|ILT-1|CD85h antigen|LIR-7|CD85H|CD85h|ILT1|LIR7|LILRA2|CD85 antigen-like family member H
- Protein/peptide name:LILRA2
- Purity:>90%
- Molecular weight:50,7 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Frequently Bought Together