Order Entry
Belgium
ContactUsLinkComponent
 
Sodium chloride solution 1 M, in single-use containers, HyClone™ for bioprocessing
 
Sodium chloride solution 1 M, in single-use containers, HyClone™ for bioprocessing

 

  • Environmentally Preferable:

 

 

  • Storage:
    Room Temperature as defined by regional Pharmacopeia (15-30°C in US; 15-25°C in EU)
  • Endotoxin Concentration:
    ≤ 1 EU/mL
  • Certifications/Compliance:
    Manufactured in a ISO9001 and ISO13485 certified facility
  • Shelf life:
    12 Months
  • For use with:
    Chromatography resins, IC systems, ÄKTA hardware systems
  • Appearance:
    Clear, colorless solution
  • Sterility:
    No Growth

 

Frequently Bought Together