Order Entry
Belgium
Orders LinkContactUsLinkComponent
Epoxy embedding medium, hardener MNA, Sigma-Aldrich®

Epoxy embedding medium, hardener MNA, Sigma-Aldrich®

Epoxy embedding medium, hardener MNA ≥95.0%, 1 L
Catalog # 45347-1L-F
Epoxy embedding medium, hardener MNA, Sigma-Aldrich®
Catalog # 45347-1L-F
CAS Number:  
Epoxy embedding medium, hardener MNA, Sigma-Aldrich®

Epoxy embedding medium, hardener MNA ≥95.0%, 1 L

CAS Number:  
Price
128,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Epoxy embedding medium is generally used for biological tissues during study by electron microscopy. Usually, it consists of three components; Epon 812 resin, an anhydride hardener and an accelerator

  • Epon™ MNA substitute

Application
It was used during transmission electron microscopy done for the analysis of epithelial cells; during a study performed to understand the epithelial cell growth and stratification in minimal culture conditions.

Epon is a trademark of Hexion, Inc.

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    Epoxy embedding medium, hardener MNA ≥95.0%

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.