2-Hydroxy-5-(pentafluorothio)benzoic acid 95+%
Catalog # APOSPC56143-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:264.17 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
CAS Number:1801700-81-1
Formula:C₇H₅F₅O₃S
MDL Number:MFCD31561289