3-Hydroxy-2-(trifluoromethyl)propanoic acid, tech
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:382-43-4
:
Formula:C₄H₅F₃O₃
MDL Number:MFCD00042314
:158.076 g/mol
Boiling Pt:96…97 °C (2.5 mm Hg)
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
Frequently Bought Together