
2-Chloro-6-(trifluoromethyl)benzylamine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:771582-27-5
:
Formula:C₈H₇ClF₃N
MDL Number:MFCD06213040
:209.596 g/mol
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
Frequently Bought Together