4-Hydroxy-2'-(trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid
Catalog # APOSPC11157-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:282.215 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:400744-89-0
Formula:C₁₄H₉F₃O₃
MDL Number:MFCD04039233