4-Hydroxy-3-iodobenzoic acid 97%
Catalog # APOSOR9769-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
Melting Pt:170…175 °C
:264.017 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:37470-46-5
Formula:C₇H₅IO₃
MDL Number:MFCD00234263