Order Entry
Belgium
ContactUsLinkComponent
5-(2-Chlorophenyl)-3-methyl-1,2,4-oxadiazole 98%
5-(2-Chlorophenyl)-3-methyl-1,2,4-oxadiazole 98%
Catalog # APOSOR960269-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(2-Chlorophenyl)-3-methyl-1,2,4-oxadiazole 98%
Catalog # APOSOR960269-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :194.618 g/mol
Storage Temperature:Ambient
CAS Number:1120271-16-0
Formula:C₉H₇ClN₂O
MDL Number:MFCD13961846