1-Methyl-DL-proline 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: N-DL-Me-Pro-OH, 1-Methylpyrrolidine-2-carboxylic acid
CAS Number:68078-09-1
:
Formula:C₆H₁₁NO₂
MDL Number:MFCD01605583
Molecular Weight:129.157 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together