
4-Propylbenzenesulphonyl chloride 96+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:146949-07-7
Formula:C₉H₁₁ClO₂S
MDL Number:MFCD00041508
:218.7 g/mol
Boiling Pt:275…276 °C
Melting Pt:20…22 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together