4-Hydroxy-2-naphthoic acid ≥98%
Catalog # APOSOR510222-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:188.179 g/mol
Storage Temperature:Ambient
CAS Number:1573-91-7
Formula:C₁₁H₈O₃
MDL Number:MFCD12405016