3-[(4-Bromophenyl)(hydroxy)methyl]isoxazole-4-carboxylic acid
Catalog # APOSOR471405-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:298.09 g/mol
Storage Temperature:Ambient
CAS Number:2006276-91-9
Formula:C₁₁H₈BrNO₄
MDL Number:MFCD28977385