N-Boc-6-hydroxy-DL-norleucine methyl ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:81505-49-9
:
Formula:C₁₂H₂₃NO₅
MDL Number:MFCD18384776
:261.315 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together