2-Hydroxy-5-isopropylbenzoic acid
Catalog # APOSOR470352-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:180.2 g/mol
Storage Temperature:Ambient
CAS Number:31589-71-6
Formula:C₁₀H₁₂O₃
MDL Number:MFCD11203639