5-Hydroxy-1H-pyrazole-3-carboxylic acid ethyl ester
Catalog # APOSOR346230-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:156.139 g/mol
Storage Temperature:Ambient
CAS Number:85230-37-1
Formula:C₆H₈N₂O₃
MDL Number:MFCD09263987