7-Bromo-4-hydroxy-8-methylquinoline-3-carboxylic acid
Catalog # APOSOR309310-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:282.09 g/mol
Storage Temperature:Ambient
CAS Number:1189107-54-7
Formula:C₁₁H₈BrNO₃
MDL Number:MFCD12674984