Order Entry
Belgium
ContactUsLinkComponent
4-Hydroxy-3-nitrobenzeneboronic acid 98%
4-Hydroxy-3-nitrobenzeneboronic acid 98%
Catalog # APOSOR1330-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Hydroxy-3-nitrobenzeneboronic acid 98%
Catalog # APOSOR1330-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:111…113 °C
 :182.927 g/mol
Storage Temperature:Refrigerator
CAS Number:850568-75-1
Formula:C₆H₆BNO₅
MDL Number:MFCD06659843