Order Entry
Belgium
Orders LinkContactUsLinkComponent
DBCO-C6-NHS ester

DBCO-C6-NHS ester

Irritant
New Product
Supplier:   Apollo Scientific
DBCO-C6-NHS ester
Supplier:  Apollo Scientific
New Product
Certificates
DBCO-C6-NHS ester

2 Options Available Below

Product Details & Documents

DBCO-C6-NHS Ester is an amine-reactive compound, which can be used to modify an amine-containing molecule in organic media. This reagent has limited solubility in aqueous media. The extended 6-carbon atom spacer arm improves solubility in commonly used organic solvents including dichloromethane, chloroform, THF, and ethyl acetate and it also improves derivatization efficiency and stability of conjugates.

Synonyms:  DBCO-NHS ester 2, Dbco-c6-nhs ester
  • CAS Number:1384870-47-6
  • Formula:C₂₅H₂₂N₂O₅
  • MDL Number:MFCD22380763
  • Molecular Weight:430.453 g/mol
  • Storage Temperature:Refrigerator
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 472,00 €
    472,00 €
    1 * 250 mg
       472,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 1 380,00 €
    1 380,00 €
    1 * 1 g
       1 380,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.