Order Entry
Belgium
Orders LinkContactUsLinkComponent
m-PEG24-NHS ester

m-PEG24-NHS ester

New Product
Supplier:   Apollo Scientific
m-PEG24-NHS ester
Supplier:  Apollo Scientific
New Product
Certificates
m-PEG24-NHS ester

3 Options Available Below

Product Details & Documents
  • CAS Number:756525-94-7
  • Formula:C₅₄H₁₀₃NO₂₈
  • MDL Number:MFCD11041155
  • Molecular Weight:1214.386 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 246,00 €
    246,00 €
    1 * 50 mg
       246,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 704,00 €
    704,00 €
    1 * 250 mg
       704,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 1 900,00 €
    1 900,00 €
    1 * 1 g
       1 900,00 €

    Specifications:
    New Product
    New Product