m-PEG16-NHS ester
New Product
Supplier: Apollo Scientific
Jump to:
About this item
- Formula:C₃₈H₇₁NO₂₀
- MDL Number:MFCD13184971
- Molecular Weight:861.965 g/mol
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
- Formula:C₃₈H₇₁NO₂₀
- MDL Number:MFCD13184971
- Molecular Weight:861.965 g/mol
- Storage Temperature:Ambient



