Order Entry
Belgium
Orders LinkContactUsLinkComponent
m-PEG3-NHS ester

m-PEG3-NHS ester

New Product
Supplier:   Apollo Scientific
m-PEG3-NHS ester
Supplier:  Apollo Scientific
New Product
Certificates
m-PEG3-NHS ester

2 Options Available Below

Product Details & Documents
  • Formula:C₁₂H₁₉NO₇
  • MDL Number:MFCD20228789
  • Molecular Weight:289.282 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 163,00 €
    163,00 €
    1 * 250 mg
       163,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 479,00 €
    479,00 €
    1 * 1 g
       479,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.