Mal-PEG6-NHS ester
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 2,5-dioxopyrrolidin-1-yl 1-(2,5-dioxo-2H-pyrrol-1(5H)-yl)-3,6,9,12,15,18-hexaoxahenicosan-21-oate
- Formula:C₂₃H₃₄N₂O₁₂
- MDL Number:MFCD20926374
- Molecular Weight:530.522 g/mol
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: 2,5-dioxopyrrolidin-1-yl 1-(2,5-dioxo-2H-pyrrol-1(5H)-yl)-3,6,9,12,15,18-hexaoxahenicosan-21-oate
- Formula:C₂₃H₃₄N₂O₁₂
- MDL Number:MFCD20926374
- Molecular Weight:530.522 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



