Mal-PEG4-NHS ester
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 2,5-Dioxopyrrolidin-1-yl 1-(2,5-dioxo-2H-pyrrol-1(5H)-yl)-3,6,9,12-tetraoxapentadecan-15-oate
- CAS Number:1325208-25-0
- Formula:C₁₉H₂₆N₂O₁₀
- MDL Number:MFCD20230464
- Molecular Weight:442.417 g/mol
- Storage Temperature:Ambient
4 Options Available Below
Product Details & Documents
Synonyms: 2,5-Dioxopyrrolidin-1-yl 1-(2,5-dioxo-2H-pyrrol-1(5H)-yl)-3,6,9,12-tetraoxapentadecan-15-oate
- CAS Number:1325208-25-0
- Formula:C₁₉H₂₆N₂O₁₀
- MDL Number:MFCD20230464
- Molecular Weight:442.417 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



