Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bis-PEG1-NHS ester

Bis-PEG1-NHS ester

New Product
Supplier:   Apollo Scientific
Bis-PEG1-NHS ester
Supplier:  Apollo Scientific
New Product
Certificates
Bis-PEG1-NHS ester

3 Options Available Below

Product Details & Documents
Synonyms:  Bis(2,5-dioxopyrrolidin-1-yl) 3,3'-oxydipropanoate
  • Formula:C₁₄H₁₆N₂O₉
  • MDL Number:MFCD20226397
  • Molecular Weight:356.285 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 225,00 €
    225,00 €
    1 * 250 mg
       225,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 568,00 €
    568,00 €
    1 * 1 g
       568,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 2 420,00 €
    2 420,00 €
    1 * 5 g
       2 420,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.