Biotin-PEG8-NHS ester
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: N-Succinimidyl 27-(Biotinamido)-4,7,10,13,16,19,22,25-octaoxaheptacosanoate, 27-(Biotinamido)-4,7,10,13,16,19,22,25-octaoxaheptacosanoic acid N-Succinimidyl ester, 2,5-Dioxopyrrolidin-1-yl 29-oxo-33-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-4,7,10,13,16,19,22,25-octaoxa-28-azatritriacontanoate
- CAS Number:2143968-03-8
- Formula:C₃₃H₅₆N₄O₁₄S
- MDL Number:MFCD27977502
- Molecular Weight:764.89 g/mol
- Storage Temperature:Ambient
4 Options Available Below
Product Details & Documents
Synonyms: N-Succinimidyl 27-(Biotinamido)-4,7,10,13,16,19,22,25-octaoxaheptacosanoate, 27-(Biotinamido)-4,7,10,13,16,19,22,25-octaoxaheptacosanoic acid N-Succinimidyl ester, 2,5-Dioxopyrrolidin-1-yl 29-oxo-33-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-4,7,10,13,16,19,22,25-octaoxa-28-azatritriacontanoate
- CAS Number:2143968-03-8
- Formula:C₃₃H₅₆N₄O₁₄S
- MDL Number:MFCD27977502
- Molecular Weight:764.89 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



