Order Entry
Belgium
ContactUsLinkComponent
Polyethylene glycol 6,000, GPR RECTAPUR®
Polyethylene glycol 6,000, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Polyethylene glycol 6,000, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Average molecular weight:
    5400 - 7000
  • Hydroxyl value:
    17 - 21
  • pH (5%):
    4,5 - 7,5
  • Heavy metals (as Pb):
    max. 10 ppm
  • Ignition residue (SO4):
    max. 0.1%
  • Water:
    max. 1.0%
  • Solidification point:
    55 - 61°C

Specification Test Results

Frequently Bought Together