Order Entry
Belgium
ContactUsLinkComponent
Tissue processor, HP300
Tissue processor, HP300
  720-2970
Tissue processor, HP300
  720-2970

 

  • Temperature Range:
    10...40 °C (operating) °C
  • Weight:
    190 kg
  • Description:
    Tissue processor, HP300 for 2 baskets, 300 cassettes
  • W×D×H:
    613×680×1320 mm
  • Electrical supply:
    220 VAC ±10%, 50/60 Hz
  • Pk:
    1

 

 

This tissue processor HP300 is a combination of unique design, built-in intelligence, and cutting-edge technology.

  • Innovative electronic locking feature and cloud platform
  • Reliability in performance
  • Safety in specimen and automatic reagent replacement
  • Smart in management (automatic infusion and smart replacement)
  • Easy in use (angle adjustable screen, liquid level observation, flexible processing capacity, easy to clean, concise UI)
  • Power-off mechanism guarantees smooth dehydration at night, safeguarding samples uninterruptedly
  • Three adjustable screen positions for flexible viewing options; 12,1" colour touch screen panel

Marble tabletop is easy to clean and serves as a thermal insulator. The air-proof inner cover has heater functionality and is resistance to the reagents and paraffin. This tissue processor incorporates an intelligent management system. Its consistent dehydration process is safe guarded by monitoring parameters for the quality of cassettes being processed, dehydration batches and days of reagent usage. The processor incorporates real-time monitoring of the running status, reagent consumption, record tracking and data analysis. Processor status can also be checked via a remote-control system. A light alarm makes it easy to check the current status, blue LED lights are designed to observe liquid level and recognise opacity conditions of reagents.

The five-point liquid level monitoring system with a liquid overflow prevention ensure through sample infusion and safety. It is equipped with a dual air pump that provides double air pressure supply with a double temperature control system enhances the performance level. The innovative liquid control system can effectively reduce impurity contamination (such as fat and paraffin), preventing pipe or valve blockage from opening.

The automatic identification of reagent and rotation is convenient and labour saving. Intelligent reagent replacing system is manual work saving; paraffin can be automatically replaced and replenished to ensure complete infusion of samples.

The double draining function of draining the paraffin before the cleaning process ensures better cleaning and saves cleaning reagents. It also has screen cleaning mode to prevent harmful interruptions during operations. There are four cleaning mode options for different user requirements.
Quick washing - without paraffin
Standard washing - with paraffin
Warm water washing - residual crystal
Fat washing - fat or tissue residual

 : Two sample container sizes are available for flexible options; contact your Avantor representative for more information.