Orders LinkContactUsLinkComponent
Molecular detection matrix control
Molecular detection matrix control, 96
Catalog # MMMPMDMC96NA
Supplier:   Neogen
Molecular detection matrix control
Catalog # MMMPMDMC96NA
Supplier:  Neogen
CAS Number:  
Molecular detection matrix control
Molecular detection matrix control
Molecular detection matrix control, 96
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 96 ST
Product Details & Documents
Kit contains 96 tests and is used with the 3M™ Molecular Detection System to check for inhibition of the matrix sample during validation periods, when adopting the 3M™ method or testing new or unknown matrices.
  • Verify that new or unknown matrices are not inhibitory to amplification
  • Ready-to-use and pre-dispensed reagents
  • Only two transfer steps after enrichment
  • Closed-tube system reduces risk of amplicon contamination
  • Designed to work with 3M™ proprietary enrichment broths to promote growth
Specifications
Specifications
  • Pk:
    96
  • Description:
    Molecular detection matrix control
Specifications