Order Entry
Belgium
ContactUsLinkComponent
Barium chloride 0,1 mol/l (0,2 N) (20°C), Titripur®, Supelco®
Barium chloride 0,1 mol/l (0,2 N) (20°C), Titripur®, Supelco®
Catalog # 1.60324.1003
Supplier:  Merck
CAS Number:  
Barium chloride 0,1 mol/l (0,2 N) (20°C), Titripur®, Supelco®
Catalog # 1.60324.1003
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Bottle with RFID tag
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :208.24 g/mol
Storage Temperature:Ambient
CAS Number:10361-37-2
Formula:BaCl₂
MDL Number:MFCD00003445