Sulphuric acid 5 mol/l (10 N), Titripur®, Supelco®
Catalog # 1.60315.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
About this item
:
:98.08 g/mol
Boiling Pt:330
UN:2796
:GHS05
Melting Pt:10.38
Storage Temperature:Ambient
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589
Density:1.84
ADR:8
:M-Clarity™ quality level = MQ100