Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium hydroxide, pellets NF, Multi-Compendial

Sodium hydroxide, pellets NF, Multi-Compendial

Corrosive
Supplier:   Avantor
Sodium hydroxide, pellets NF, Multi-Compendial
Supplier:  Avantor
Certificates
Sodium hydroxide, pellets NF, Multi-Compendial
Sodium hydroxide, pellets NF, Multi-Compendial

2 Options Available Below

Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:1390 °C
  • Melting Pt:323 °C
  • Density:2.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • UN:1823
  • ADR:8,II

Documents

Specifications for Product Family

Specification Test Results

  • NF - Identification A (Sodium):
    Passes Test
  • NF - Identification B (pH):
    ≥11.0
  • NF - Insoluble Substances and Organic Matter:
    Passes Test
  • NF - Potassium (K):
    ≤0.5 %
  • NF - Carbonate (as Na₂CO₃):
    ≤3.0 %
  • NF - Total Alkali:
    95.0 - 100.5 %
  • NF - Content of Sodium:
    54.0 - 59.8 %
  • ChP -Assay:
    97.0 - 100.5 %
  • ChP -Sodium carbonate:
    2.0% max
  • ChP -Identification:
    Passes Test
  • ChP -Alkalinity:
    Passes Test
  • ChP -Clarity and color of solution :
    Passes Test
  • ChP -Chloride :
    0.005 % max
  • ChP -Sulfate :
    0.015 % max
  • ChP -Potassium :
    Passes Test
  • ChP -Aluminum and iron :
     0.1% max
  • ChP -Iron :
    0.001% max
  • ChP -Heavy metal :
    20 ppm max
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Carbonate (as Na₂CO₃):
    ≤2.0 %
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤200 ppm
  • EP/BP - Sulfate (SO₄):
    ≤200 ppm
  • EP/BP - Iron (Fe):
    ≤10 ppm
  • EP/BP - Assay (NaOH):
    97.0 - 100.5 %
  • JP - Assay:
    95.0 - 101.0 %
  • JP - Identification A:
    Passes Test
  • JP - Identification B:
    Passes Test
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Chloride (Cl):
    ≤0.050 %
  • JP - Heavy Metals (as Pb):
    ≤30 ppm
  • JP - Potassium (K):
    Passes Test
  • JP - Carbonate (as Na₂CO₃):
    ≤2.0 %
  • JP - Mercury (Hg):
    Passes Test
  • Endotoxin Concentration (EU/g):
    ≤5
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Poly bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 158,70 €
    158,70 €
    1 * 500 g
       158,70 €

  • PE pail, screw top
    12 kg
    Log in for availability
    Log in for availability
    1 * 12 kg
    1 * 12 kg - 2 066,35 €
    2 066,35 €
    1 * 12 kg
       2 066,35 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.