ContactUsLinkComponent
Control slide, Melanin TISSUE-TROL™, Sigma-Aldrich®
Control slide, Melanin TISSUE-TROL™, Sigma-Aldrich®
  TTR014-25EA
Control slide, Melanin TISSUE-TROL™, Sigma-Aldrich®
  TTR014-25EA
  • Description:
    Melanin TISSUE-TROL™ control slide
  • Pk:
    25
Histology slides with paraffin embedded section of human skin containing melanin.
Suitable for use with Fontana-Masson Stain Kit (Procedure No. HT200).