ContactUsLinkComponent
Control slide, Fungi TISSUE-TROL™, Sigma-Aldrich®
Control slide, Fungi TISSUE-TROL™, Sigma-Aldrich®
Catalog # TTR004-25EA
undefined
Control slide, Fungi TISSUE-TROL™, Sigma-Aldrich®
Catalog # TTR004-25EA
Specifications
  • Description:
    Fungi TISSUE-TROL™ control slide
  • Pk:
    25
Specifications
About this item
Histology slides with paraffin embedded section of mouse lung tissue containing Candida albicans.
Suitable for use with Silver Stain (Modified GMS) Procedure No. HT100.