
Procion® Red MX-5B, Sigma-Aldrich®
Catalog # 404365-5G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:615.33 g/mol
Melting Pt:>300 °C (lit.)
:GHS08
Storage Temperature:Ambient
CAS Number:17804-49-8
Formula:C₁₉HCl₂N₆Na₂O₇S₂
MDL Number:MFCD00067094